Fiberlite™ F8-6 x 1000y Fixed-Angle Large Capacity Rotor
Thermo Scientific™
Fiberlite™ F8-6 x 1000y Fixed-Angle Large Capacity Rotor
Get high throughput and performance from a fixed-angle rotor that supports up to six 1L bottles in a single spin while operating up to 8,500rpm and 15,860 x g.
Have Questions?
Catalog NumberCapacity
096-0611376 x 1000 mL
Catalog number 096-061137
Price (USD)
-
Capacity:
6 x 1000 mL
Experience high throughput and high performance from the Thermo Scientific™ Fiberlite™ F8-6x1000y Fixed-Angle Large Capacity Rotor, which supports up to six 1L bottles in a single spin while operating up to 8,500rpm and 15,860 x g.
Fiberlite rotors:
  • Lightweight carbon fiber construction reduces run-times with higher g-forces.
  • Corrosion-and fatigue-resistant design ensures structural integrity.
  • Rotors are backed by a 15-year warranty
Ordering information:
Order rotor body (096-061137) for complete assembly. For refresher kit, order 021-061137; Lid assembly, order 099-061137; replacement O-rings for lids, order 022-061137 separately. Contact your sales representative for information.
Compatible with:
Thermo Scientific™ Sorvall™ RC BIOS, Nalgene™ bottles and tubes
Specifications
Angle20°
DescriptionFiberlite F8-6 x 1000y Fixed-Angle Large Capacity Rotor
For Use With (Equipment)Thermo Scientific Sorvall RC BIOS 6 Large Capacity Centrifuges
K Factor5096
Max. Tube Diameter98 mm
Max. Tube Length195 mm
Min. RCF4,369 x g
Radius (Metric)196 mm
Radius (Metric) Min.54 mm
Capacity6 x 1000 mL
MaterialCarbon Fiber Composite
Max. RCF15,900 x g
Max. Speed8,500 rpm
TypeFixed Angle Rotor
Unit SizeEach