DRI™Serum Toxicology Calibrator 4
Product Image
Thermo Scientific™
DRI™Serum Toxicology Calibrator 4
Use Thermo Scientific™ DRI™ Serum Toxicology Calibrator 4 with Serum Toxicology Barbiturate, Serum Toxicology Benzodiazepine and Tricyclics immunoassays.
Have Questions?
Catalog NumberQuantity
09765 mL
Catalog number 0976
Price (USD)
-
Request A Quote
Quantity:
5 mL
Use Thermo Scientific™ DRI™ Serum Toxicology Calibrator 4 with Serum Toxicology Barbiturate, Serum Toxicology Benzodiazepine and Tricyclics immunoassays. Liquid, ready-to-use calibrators are conveniently packaged for use with serum toxicology immunoassays.
  • Multianalyte calibrators
Ordering Alerts:
0976: 1 Bottle, 5mL. Concentrations: Barbiturates 5000ng/mL, Benzodiazepine 200ng/mL and Tricyclics 1000ng/mL.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSerum Toxicology Calibrator 4
Quantity5 mL
TypeCalibrator
Detectable AnalytesBarbiturate, Benzodiazepines, Tricyclic Antidepressants, Serum Barbiturate, Serum Benzodiazepine, Serum Tricyclic
Sample TypeSerum
Unit SizeEach
Contents & Storage
2°C to 8°C