HAAKE™ RheoWin™ Measuring and Evaluation Software
Actual product may vary
HAAKE™ RheoWin™ Measuring and Evaluation Software
HAAKE™ RheoWin™ Measuring and Evaluation Software
HAAKE™ RheoWin™ Measuring and Evaluation Software
HAAKE™ RheoWin™ Measuring and Evaluation Software
HAAKE™ RheoWin™ Measuring and Evaluation Software
HAAKE™ RheoWin™ Measuring and Evaluation Software
HAAKE™ RheoWin™ Measuring and Evaluation Software
Thermo Scientific™
HAAKE™ RheoWin™ Measuring and Evaluation Software
Easily navigate your rheometer or viscometer operation using RheoWin Measuring and Evaluation Software.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호유형용도(장비)용도(애플리케이션)
098-5080SoftwareHAAKE MARS iQ RheometerNavigating System Operations
098-5075SoftwareHAAKE Viscotester iQ AirNavigating System Operations
098-5077SoftwareHAAKE MARS iQ Air RheometerNavigating System Operations
098-5072SoftwareHAAKE MARS 40 RheometerNavigating System Operations
098-5071SoftwareHAAKE MARS 60 RheometerNavigating System Operations
098-5566Software ModuleHAAKE RheoWin SoftwareAsphalt/Bitumen
098-5045Software ModuleHAAKE RheoWin SoftwareTime Temperature Superposition
098-5044Software ModuleHAAKE RheoWin Software21 CFR part 11 tool
098-5057Software ModuleHAAKE RheoWin SoftwareInterfacial Rheology
098-5061Software ModuleHAAKE RheoWin SoftwareFT-IR Spectroscopy
098-5062SoftwareHAAKE Viscotester iQNavigating System Operations
카탈로그 번호 098-5080
제품 가격(KRW)
-
유형:
Software
용도(장비):
HAAKE MARS iQ Rheometer
용도(애플리케이션):
Navigating System Operations
Easily navigate your rheometer or viscometer operation using the Thermo Scientific™ HAAKE™ RheoWin™ Measuring and Evaluation Software. This user-friendly software for rheometers and viscometers can be customized to offer convenient rheometer or viscometer operation with default settings for beginners and full access to all relevant measuring parameters for advanced rheologists.
The HAAKE RheoWin Measuring and Evaluation Software Features
Customizable Software
  • RheoWin JobManager for fully automated process control ('jobs') of measuring and analysis routines and report printout or export
  • RheoWin DataManager for interactive evaluation of measured data, as well as sophisticated tools for creating reports and generating templates for graphs, tables and screen views
  • RheoWin UserManager for comprehensive management of user access control and assignment of specific access rights
Functionality
  • Manual control for preliminary testing, selected parameters display and saving manually-acquired data
  • Convenient creation and customization of measuring routines using predefined measuring and evaluation elements via 'drag and drop' techniques
  • Fully automated measuring, data evaluation and documentation within one measuring routine
  • Simultaneous measurements using several instruments and data evaluation
  • Freely configurable data export (e.g., ASCII, MS-Excel, XML, etc.)
  • Ability to save graphs in various formats
  • Numerous algorithms for data analysis (e.g., interpolation, regression and automated quality control)
  • Ability to save raw data and numerical values for quality control purposes
  • Integrated image capture with USB and firewire camera
Customization
  • User-defined configuration of paths and subdirectories for data filing
  • Modular generation of a file name and automated saving in a predefined subdirectory
  • Data transfer to ERP and laboratory systems (e.g., SAP™, LIMS, etc.)
  • Online display of all corrections to improve data quality
  • Snapshot for quick characterization of an unknown sample
사양
용도(애플리케이션)Navigating System Operations
용도(장비)HAAKE MARS iQ Rheometer
유형Software
Unit SizeEach