Product Image
Thermo Scientific™

DRI™ Ecstasy Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
100081500 ng/mL
1000791000 ng/mL
100080750 ng/mL
100082250 ng/mL
Catalog number 100081
Price (USD)
-
Concentration:
500 ng/mL

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration500 ng/mL
DescriptionDRI Ecstasy Drugs of Abuse Calibrators
Packaging1 Bottle
Quantity1 x 10 mL
TypeCalibrator
Detectable AnalytesMDA, MDEA, MDMA
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C