PBS, pH 7.4
Gibco™

PBS, pH 7.4

Gibco PBS, pH 7.4, is a balanced salt solution ideal for cell culture applications. It lacks calcium, magnesium, and phenol red, preventing cell adhesion and assay interference. Trusted for quality, Gibco PBS ensures reliable results in cell washing, transport, and reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1001004910 x 500 mL
100100311000 mL
10010023500 mL
100100726 x 1000 mL
Catalog number 10010049
Price (USD)
250.65
Online exclusive
298.00
Save 47.35
Each
Quantity:
10 x 500 mL
Price (USD)
250.65
Online exclusive
298.00
Save 47.35
Each

Gibco Phosphate Buffered Saline (PBS), pH 7.4, is a balanced salt solution widely used in various cell culture applications. This product is specifically formulated without calcium, magnesium, and phenol red, making it ideal for a range of laboratory procedures.

Key features

  • Without calcium and magnesium—the absence of calcium and magnesium prevents unwanted cell adhesion and aggregation, which is crucial during cell dissociation and flow cytometry
  • Without phenol red—the lack of phenol red ensures that there is no interference with optical measurements and prevents potential effects on cell physiology

Applications

  • Washing cells before dissociation—PBS is commonly used to rinse cells before enzymatic dissociation, ensuring that chelators are removed from the culture
  • Transporting cells or tissue—PBS provides a stable environment for cells or tissues during transportation
  • Diluting cells for counting—PBS is used to dilute cell suspensions for accurate counting
  • Preparing reagents—PBS serves as a buffer in the preparation of various reagents

Why choose PBS over DPBS

  • Simplicity—PBS is a simpler formulation compared to Dulbecco's Phosphate-Buffered Saline (DPBS), which includes additional components like potassium chloride and may be formulated with or without calcium, magnesium, glucose, and pyruvate. This simplicity makes PBS a versatile choice for general cell culture applications.
  • Specific applications—PBS is preferred for applications where the presence of potassium or other additives in DPBS might interfere with experimental outcomes

Why choose pH 7.4 over pH 7.2

  • Physiological relevance—pH 7.4 is closer to the physiological pH of human blood, making it more suitable for maintaining the natural state of cells during experiments. This pH is particularly important for applications involving mammalian cells.

Why choose Gibco PBS, pH 7.4

  • Quality and reliability—manufacture of Gibco PBS, pH 7.4, is cGMP-compliant under the ISO 13485 standard, ensuring high standards of quality and consistency
  • Extensive testing—lot release testing of osmolality, pH levels, stability, endotoxins, and sterility ensures the quality of the product
  • Proven track record—Gibco cell culture products have a long-standing reputation in the scientific community, with numerous publications citing the use of Gibco PBS, reflecting its widespread acceptance and trust among researchers

Gibco PBS, pH 7.4, is a reliable and high-quality solution for various applications. Its formulation without calcium, magnesium, and phenol red makes it versatile and suitable for specific experimental needs. Choosing Gibco PBS ensures consistency, quality, and support from a trusted brand in the scientific community.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialPhosphate Buffered Saline (PBS)
ColorClear
IncludesNo Calcium, No Magnesium
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality280 to 315 mOsm/kg
PackagingBottle
Recommended StorageStorage conditions: 15°C to 30°C, Shipping conditions: Room temperature, Shelf life: 24 months from date of manufacture
SterilitySterile
Sterilization MethodSterile-filtered
Concentration1X
For Use With (Application)Washing and Staining Cells
GradeCell Culture
Physical FormLiquid
Product LineGibco
Quantity10 x 500 mL
Solution TypepH Buffer, Storage Solution and Cleaning Solution
pH7.4
Unit SizeEach

Frequently asked questions (FAQs)

My PBS, PH 7.4 (Cat. No. 10010072) was accidentally frozen. Can I still use it?

This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products tested for endotoxin levels?

Our PBS, pH 7.2 and PBS, pH 7.4 products are tested for endotoxin and the endotoxin levels are shown on the Certificate of Analysis (CoA) for the particular lot number of the product.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products sterile and suitable for cell culture?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625), which is molecular biology grade and is certified to be RNase-free. However, it is not sterile and hence is not cell culture-grade.

All our other PBS products (Cat. Nos. 70011xxx, 10010xxx, 70013xxx, 20012xxx) are filter-sterilized using a 0.1 µm filter and are cell culture grade (ready to use directly on cells). However, they are not tested for nuclease activity.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your PBS products DNase- and RNase-free?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625) that is molecular biology grade and is certified to be RNase-free. Our other PBS products are made with distilled water and are filter-sterilized, but are not tested for RNase and DNase activity.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
A 2-D liquid separations/mass mapping method for interlysate comparison of ovarian cancers.
Authors: Kachman Maureen T; Wang Haixing; Schwartz Donald R; Cho Kathleen R; Lubman David M;
Journal:Anal Chem
PubMed ID:11985308
A two-dimensional liquid phase separation of proteins from whole cell lysates coupled on-line to an electrospray-ionization time-of-flight (ESI-TOF) mass spectrometer (MS) is used to map the protein content of ovarian surface epithelial cells (OSE) and an ovarian carcinoma-derived cell line (ES2). The two dimensions involve the use of liquid isoelectric ... More