Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer
Thermo Scientific™
Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
10011401Smart Rotary Cell Changer BaseTemperature Probe
840-214900Evolution Spectrophotometers7-Cell Rotary Changer
10010110Smart Rotary Cell Changer BaseSpare 7-cell Rotary Changer
Catalog number 10011401
Price (USD)
-
For Use With (Equipment):
Smart Rotary Cell Changer Base
Type:
Temperature Probe
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer. Combined with a liquid recirculator, this cell changer provides control of sample temperature for up to 7 samples or standards.
  • Easy-to-use sample positioning accessory for use with Thermo Scientific™ Evolution™ One/One Plus and Evolution™ Pro spectrophotometers.
  • Accessory control fully integrated in Thermo Scientific™ INSIGHT™ Pro software.
  • Automatic software recognition, smart alignment and serial number reporting
  • Includes carousel and motor base.
  • When placed in both sample and reference beams (Evolution 300 only), this cell changer allows comparison of up to 7 samples with 7 unique references.
  • Monitor the temperature in a cell with Temperature probe accessory for Smart accessories, catalog number 0011401.
  • When deploying this accessory in a regulated or QC environment, a full-system qualification package is available to assure compliance of instrument and accessory as a complete unit.
Specifications
Configuration DetailsSingle temperature probe. Connects directly to the Smart accessory motor base. Monitors the temperature in one cuvette.
For Use With (Equipment)Smart Rotary Cell Changer Base
TypeTemperature Probe
Unit SizeEach