Attune™ Xenith™ 349 nm Laser Line Filters
Invitrogen™

Attune™ Xenith™ 349 nm Laser Line Filters

These replacement optical filters are for those channels available off the 349 nm laser of the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeExcitation/Emission
100117854CSDichroic Shortpass Filter560DSP
100117801CSEmission Filter670/30
100117802CSEmission Filter728/40
100117803CSEmission Filter810/25
100117804CSEmission Filter615/24
100117805CSEmission Filter575/15
100117806CSEmission Filter549/15
100117807CSEmission Filter507/19
100117808CSEmission Filter387/11
100117809CSEmission Filter420/10
100117810CSEmission Filter455/14
100117811CSEmission Filter434/17
100117812CSEmission Filter473/15
100117852CSDichroic Longpass Filter530DLP
100117853CSDichroic Longpass Filter600DLP
100117855CSDichroic Longpass Filter750DLP
100117856CSDichroic Longpass Filter685DLP
100117857CSDichroic Shortpass Filter640DSP
100117858CSMirrorNot Available
100117859CSDichroic Longpass Filter409DLP
100117860CSDichroic Longpass Filter488DLP
100117861CSDichroic Longpass Filter427DLP
100117862CSDichroic Shortpass Filter465DSP
100117863CSDichroic Longpass Filter445DLP
Catalog number 100117854CS
Price (USD)
162.00
Each
Product Type:
Dichroic Shortpass Filter
Excitation/Emission:
560DSP
Price (USD)
162.00
Each

The Attune Xenith Flow Cytometer is designed with a flexible optical deck and the ability to run both spectral unmixing and conventional compensation assays on one system. Part of this flexibility is delivered through user-changeable optical filters. These optical filters are for the channels available off the 349 nm laser.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
ColorUV
Excitation/Emission560DSP
Quantity1 Ea.
Wavelength349 nm
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeDichroic Shortpass Filter
Unit SizeEach