CEDIA™ Oral Fluid Drugs of Abuse Assays
Actual product may vary
Thermo Scientific™
CEDIA™ Oral Fluid Drugs of Abuse Assays
Accurately detect drugs of abuse in oral fluid specimens with Thermo Scientific™ CEDIA™ Oral Fluid Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
10014947CEDIA Amphetamine OFT Assay
10014910CEDIA Cannabinoids OFT Assay
10014764CEDIA Cocaine OFT Assay
10014949CEDIA Methamphetamine OFT Assay
10014873CEDIA Opiate OFT Assay
10014888CEDIA Phencyclidine (PCP) OFT Assay
Catalog number 10014947
Price (USD)
-
Description:
CEDIA Amphetamine OFT Assay
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Oral Fluid Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsCEDIA Multi-Drug OFT
DescriptionCEDIA Amphetamine OFT Assay
Quantity1 x 65 mL
Sample TypeOral Fluid
TypeReagent
Detectable AnalytesAmphetamine (AMP)
DoA CalibratorsMulti-drug OFT
Unit SizeEach
Contents & Storage
2°C to 8°C