CEDIA™ Oral Fluid Drugs of Abuse Calibrators and Controls
Actual product may vary
Thermo Scientific™

CEDIA™ Oral Fluid Drugs of Abuse Calibrators and Controls

Eliminate time-consuming steps, yet achieve high analytical accuracy with the ready-to-use Thermo Scientific™ CEDIA™ Oral Fluid Drugs of Abuse Calibrators and Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityTypeDetectable Analytes
100149541 x 20 mLNegative CalibratorAmphetamine (AMP), Benzodiazepines (BZO), Cocaine (COC), Morphine, Opiates (OPI), Phencyclidine (PCP)
100149551 x 10 mLCutoff CalibratorAmphetamine (AMP), Benzodiazepines (BZO), Cocaine (COC), Morphine, Opiates (OPI), Phencyclidine (PCP)
100149572 x 15 mLControl Set (Low and High)Amphetamine (AMP), Benzodiazepines (BZO), Cocaine (COC), Morphine, Opiates (OPI), Phencyclidine (PCP)
100149501 x 10 mLNegative CalibratorMethamphetamine (mAMP)
100149511 x 5 mLCutoff CalibratorMethamphetamine (mAMP)
100149532 x 10 mLControl Set (Low and High)Methamphetamine (mAMP)
100149221 x 10 mLNegative CalibratorCannabinoids (THC)
100149231 x 5 mLCutoff CalibratorCannabinoids (THC)
100149252 x 10 mLControl Set (Low and High)Cannabinoids (THC)
Catalog number 10014954
Price (USD)
-
Quantity:
1 x 20 mL
Type:
Negative Calibrator
Detectable Analytes:
Amphetamine (AMP), Benzodiazepines (BZO), Cocaine (COC), Morphine, Opiates (OPI), Phencyclidine (PCP)
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Oral Fluid Drugs of Abuse Calibrators and Controls, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and by developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration0 ng/mL
IncludesAmphetamine: 0 ng/mL; Cocaine: 0 ng/mL; Opiates: 0 ng/mL; Phencyclidine: 0 ng/mL
Quantity1 x 20 mL
Sample TypeOral Fluid
TypeNegative Calibrator
Detectable AnalytesAmphetamine (AMP), Benzodiazepines (BZO), Cocaine (COC), Morphine, Opiates (OPI), Phencyclidine (PCP)
Unit SizeEach
Contents & Storage
2°C to 8°C