Product Image
Thermo Scientific™
CEDIA™ Sample Check Assay
A type of specimen validity test that is used to determine if the urine specimen contains any compounds that will compromise the validity of the urine sample using the CEDIA assays for drugs of abuse screening.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1001644385 mL, 8 x 18 mL Indiko vials
181555585 mL
Catalog number 10016443
Price (USD)
-
Quantity:
85 mL, 8 x 18 mL Indiko vials
The CEDIA™ Sample Check Assay is a type of specimen validity test that is used to determine if the urine specimen contains any compounds that will compromise the validity of the CEDIA assays for drugs of abuse screening. In order to determine the specific interfering substance(s), more specific specimen validity tests should be used, such as the DRI™ pH-Detect™ Test, the DRI™ Creatinine-Detect™ Test, the DRI™ General Oxidant-Detect™ Test, and the DRI™ Gravity-Detect™ Test.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSample check specimen validity test
Quantity85 mL, 8 x 18 mL Indiko vials
Sample TypeUrine
Shelf LifeUntil Label Expiry Date
TypeReagent
Unit SizeEach
Contents & Storage
2°C to 8°C