DRI™ Hydrocodone Drugs of Abuse Calibrators
Actual product may vary
Thermo Scientific™
DRI™ Hydrocodone Drugs of Abuse Calibrators
Save time while maintaining accuracy with the liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
10018080300 ng/mL
10018079100 ng/mL
10018081500 ng/mL
100180821000 ng/mL
Catalog number 10018080
Price (USD)
-
Concentration:
300 ng/mL
Ensure accuracy and precision of your drug testing results by partnering these liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators with our Thermo Scientific DRI Hydrocodone Drugs of Abuse Assay and Control Set.These calibrators are for sale in the U.S.A. and Canada only.
  • Specific detection of hydrocodone
  • Qualitative or semi-quantitative modes
  • For sale in the U.S.A. and Canada only
Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details
For In Vitro Diagnostic Use.  
Specifications
Concentration300 ng/mL
DescriptionDRI Hydrocodone Drugs of Abuse Calibrator
Packaging1 Bottle
Quantity1 x 10 mL
TypeCalibrator
Detectable AnalytesHydrocodone
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C