Product Image
Thermo Scientific™

DRI™ Propoxyphene Drugs of Abuse Assay

Eliminate time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Propoxyphene Drugs of Abuse Assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100185103 x 18 mL
04321 x 100 mL
04331 x 500 mL
Catalog number 10018510
Price (USD)
-
Quantity:
3 x 18 mL
Eliminate time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Propoxyphene Drugs of Abuse Assay. This liquid, ready-to-use assay delivers high analytical accuracy for an array of clinical chemistry analyzers. This liquid, ready-to-use assay has been field-tested on a wide range of analyzers. It is enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Quantity3 x 18 mL
Sample TypeUrine
Shelf LifeUntil Label Expiry Date
TypeReagent
Detectable AnalytesPropoxyphene (PPX)
Unit SizeEach
Contents & Storage
2°C to 8°C