CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls
Actual product may vary
Thermo Scientific™
CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls
Monitor assay performance with the Thermo Scientific™ CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
10020802CEDIA Buprenorphine II Calibrator 100ng/mL
10020800CEDIA Buprenorphine II Calibrator 20ng/mL
10020799CEDIA Buprenorphine II 10ng/mL Cutoff Calibrator
10021390CEDIA Negative Calibrator II
10020804CEDIA Buprenorphine II Control Set: Low (7.5ng/mL), High (12.5ng/mL)
10020801CEDIA Buprenorphine II Calibrator 50ng/mL
Catalog number 10020802
Price (USD)
-
Description:
CEDIA Buprenorphine II Calibrator 100ng/mL
Achieve qualitative and semi-quantitative test results with the Thermo Scientific™ CEDIA™ Buprenorphine II Calibrators and Controls. The five calibrators, including negative, are used to establish a standard curve that correlates the reaction signals from the analyzer with the calibrators’ respective concentrations (in ng/mL). Two controls, one above and one below the cutoff (10ng/mL), validate the calibration curve. The calibrators and controls also provide a reference point for qualitative (positive or negative results) and semi-quantitative detection.
  • Ensures assay accuracy
  • 18 months shelf life
  • Supports qualitative and semi-quantitative measurements
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration100 ng/mL
DescriptionCEDIA Buprenorphine II Calibrator 100ng/mL
Quantity1 x 5 mL
TypeCalibrator
Detectable AnalytesBuprenorphine
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C