CEDIA™ UR-144/XLR-11 (Synthetic Cannabinoids) Drugs of Abuse Assay (CJ&F)
Actual product may vary
Thermo Scientific™
CEDIA™ UR-144/XLR-11 (Synthetic Cannabinoids) Drugs of Abuse Assay (CJ&F)
Determine the presence of UR-144/XLR-11 and their metabolites in human urine with this broad- spectrum synthetic cannabinoid immunoassay. CJ&F only.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100229551 x 65 mL
100229493 x 17 mL
Catalog number 10022955
Price (USD)
-
Quantity:
1 x 65 mL
Detect UR-144 and XLR-11 (fluorinated form of UR-144: 5-F-UR144) and their metabolites with the Thermo Scientific™ CEDIA™ UR-144/XLR-11 (Synthetic Cannabinoids) Drugs of Abuse Assay (CJ&F), a broad- spectrum, synthetic cannabinoid assay. It broadly cross-reacts with 15 structurally similar derivatives, allowing you to detect as many variants as possible within a single assay. Semi-quantitative option allows you to identify borderline samples below the cutoff that might be positive (confirmed with LC-MS/MS). For Criminal Justice and Forensic use only in the USA.
  • A synthetic cannabinoid assay that specifically detects UR-144 and XLR-11 (5-F-UR144)
  • Cross-reacts with 15 derivatives, including XLR-12, AM-2201 N-(4-hydroxypentyl), JWH-250 N-(5-hydroxypentyl), JWH-018 N-pentanoic acid, and AKB-48 N-pentanoic acid
  • May detect other synthetic cannabinoids due to cumulative response
  • Good correlation to LC-MS/MS
  • Qualitative and semi-quantitative detection
  • Applications available on a wide range of clinical analyzers
For Criminal Justice and Forensic Use Only.
For Criminal Justice and Forensic Use Only.
Specifications
Control SetsCEDIA UR-144 Control Set
DescriptionCEDIA UR-14/XLR-11 Assay
Quantity1 x 65 mL
TypeReagent
Detectable AnalytesUR-144, XLR-11
DoA CalibratorsUR-144 Calibrator, Negative Calibrator II
Unit SizeEach
Contents & Storage
2°C to 8°C