CEDIA™ AB-PINACA (Synthetic Cannabinoid) Drugs of Abuse Assay (CJ&F)
Actual product may vary
Thermo Scientific™

CEDIA™ AB-PINACA (Synthetic Cannabinoid) Drugs of Abuse Assay (CJ&F)

Determine the presence of AB-PINACA, its fluorinated form, and its major metabolites with this broad-spectrum synthetic cannabinoid immunoassay. CJ&F only.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100229771 x 65 mL
100229713 x 17 mL
Catalog number 10022977
Price (USD)
-
Quantity:
1 x 65 mL
Specifically detect AB-PINACA, its fluorinated form (5-F-AB-PINACA), and its major metabolites with the Thermo Scientific™ CEDIA™ AB-PINACA (Synthetic Cannabinoid) Drugs of Abuse Assay (CJ&F), a broad- spectrum, synthetic cannabinoid assay. It broadly cross-reacts with 34 structurally similar derivatives, allowing you to detect as many variants as possible within a single assay, including AB-CHMINACA, AB-FUBINACA, AB-CHMICA, ADBICA, 5-F-ABICA, and APP-CHMINACA. Semi-quantitative option allows you to identify borderline samples below the cutoff that might be positive (confirmed by LC-MS/MS).
  • Specifically detects AB-PINACA, its fluorinated form (5-F-AB-PINACA), and five metabolites
  • Cross-reacts with 34 derivatives, including AB-CHMINACA, AB-FUBINACA, AB-CHMICA, ADBICA, 5-F-ABICA, and APP-CHMINACA
  • Good correlation to LC-MS/MS
  • Qualitative and semi-quantitative detection
  • May detect other synthetic cannabinoids due to cumulative response
  • Applications available on a wide range of clinical analyzers
For Criminal Justice and Forensic Use Only.
For Criminal Justice and Forensic Use Only.
Specifications
Control SetsCEDIA AB-PINACA Control Set
DescriptionCEDIA AB-PINACA Assay
Quantity1 x 65 mL
TypeReagent
Detectable AnalytesAB-PINACA
DoA CalibratorsAB-PINACA Calibrator, Negative Calibrator III
Unit SizeEach
Contents & Storage
2°C to 8°C