AcroMetrix™ Solid Tumor Controls (RUO)
AcroMetrix™ Solid Tumor Controls (RUO)
AcroMetrix™ Solid Tumor Controls (RUO)
Thermo Scientific™
AcroMetrix™ Solid Tumor Controls (RUO)
Validate and assess run-to-run assay variation using the Thermo Scientific™ AcroMetrix Solid Tumor Controls (RUO).
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호유형
10024431AcroMetrix Core Hotspot-CNV DNA Control (RUO)
10024432AcroMetrix Core Fusion RNA Control (RUO)
10026229AcroMetrix Hotspot Frequency Ladder (RUO)
카탈로그 번호 10024431
제품 가격(KRW)
-
유형:
AcroMetrix Core Hotspot-CNV DNA Control (RUO)
Validate and assess run-to-run solid tumor assay variation using Thermo Scientific AcroMetrix Solid Tumor Controls (RUO). These controls are intended for use with next generation sequencing (NGS), qualitative and quantitative PCR research assays that are designed to identify fusions, CNVs, SNVs and indels. These controls are for research use only.
AcroMetrix Core Fusion RNA Control (RUO)
  • Includes SLC34A2-ROS1 & EML4-ALK fusions
AcroMetrix Core Hotspot-CNV DNA Control (RUO)
  • Five hotspots and 1 CNV representative across various cancers
AcroMetrix Hotspot Frequency Ladder (RUO)
  • 555 hotspots include 504 SNV, 2 MNV, 29 DEL, 19 INS and 1 complex indel all in one control at 50%, 25%, 15%, 10%, 5%, 2.5% variant allele frequency
  • Includes a negative control at 0% variant allele frequency
연구전용. 진단용으로 사용할 수 없음.
연구전용. 진단용으로 사용할 수 없음.
사양
포장8 Vials/Kit, 8 μL/Vial
수량1 Kit
유형AcroMetrix Core Hotspot-CNV DNA Control (RUO)
Unit SizeEach
구성 및 보관
-20°C