DRI™ Oxycodone Drugs of Abuse Assays
Thermo Scientific™
DRI™ Oxycodone Drugs of Abuse Assays
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1002481 x 68 mL
1002491 x 500 mL
100156323 x 18 mL
Catalog number 100248
Price (USD)
-
Quantity:
1 x 68 mL
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs. Supports new SAMHSA guidelines (effective October 1, 2017)
  • Supports new SAMHSA guidelines (effective October 1, 2017)
  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsDRI Oxycodone Control Set
DescriptionDRI Oxycodone Drugs of Abuse Assays
Quantity1 x 68 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesOxycodone (OXY)
DoA CalibratorsOxycodone Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C