AcroMetrix™ Solid Tumor Controls (RUO)
Thermo Scientific™
AcroMetrix™ Solid Tumor Controls (RUO)
Validate and assess run-to-run assay variation using the Thermo Scientific™ AcroMetrix Solid Tumor Controls (RUO).
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerTyp
10026229AcroMetrix Hotspot Frequency Ladder (RUO)
10024431AcroMetrix Core Hotspot-CNV DNA Control (RUO)
10024432AcroMetrix Core Fusion RNA Control (RUO)
Katalognummer 10026229
Preis (EUR)
-
Typ:
AcroMetrix Hotspot Frequency Ladder (RUO)
Validate and assess run-to-run solid tumor assay variation using Thermo Scientific AcroMetrix Solid Tumor Controls (RUO). These controls are intended for use with next generation sequencing (NGS), qualitative and quantitative PCR research assays that are designed to identify fusions, CNVs, SNVs and indels. These controls are for research use only.
AcroMetrix Core Fusion RNA Control (RUO)
  • Includes SLC34A2-ROS1 & EML4-ALK fusions
AcroMetrix Core Hotspot-CNV DNA Control (RUO)
  • Five hotspots and 1 CNV representative across various cancers
AcroMetrix Hotspot Frequency Ladder (RUO)
  • 555 hotspots include 504 SNV, 2 MNV, 29 DEL, 19 INS and 1 complex indel all in one control at 50%, 25%, 15%, 10%, 5%, 2.5% variant allele frequency
  • Includes a negative control at 0% variant allele frequency
Ausschliesslich zu Forschungszwecken. Nicht zugelassen für diagnostische Verfahren.
Ausschliesslich zu Forschungszwecken. Nicht zugelassen für diagnostische Verfahren.
Specifications
Verpackung7 Vials/Kit, 30 μL/Vial
Menge1 Kit
TypAcroMetrix Hotspot Frequency Ladder (RUO)
Unit SizeEach
Inhalt und Lagerung
-20 °C