Search
Search

| Catalog Number | Quantity | Type | Concentration |
|---|---|---|---|
| 10026590 | 1 x 5 mL | Calibrator | 20 ng/mL |
| 10026591 | 1 x 5 mL | Calibrator | 50 ng/mL |
| 10026592 | 1 x 5 mL | Calibrator | 100 ng/mL |
| 10026593 | 1 x 5 mL | Calibrator | 200 ng/mL |
| 10026594 | 2 x 5 mL | Control | Low 37.5 ng/mL, High 62.5 ng/mL |
Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific CEDIA Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.