CEDIA™ Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F)
Thermo Scientific™
CEDIA™ Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F)
Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay using the Thermo Scientific CEDIA Mitragynine (Kratom) Drugs of Abuse Calibrators and Controls (CJ&F only).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityTypeConcentration
100265911 x 5 mLCalibrator50 ng/mL
100265901 x 5 mLCalibrator20 ng/mL
100265921 x 5 mLCalibrator100 ng/mL
100265931 x 5 mLCalibrator200 ng/mL
100265942 x 5 mLControlLow 37.5 ng/mL, High 62.5 ng/mL
Catalog number 10026591
Price (USD)
-
Quantity:
1 x 5 mL
Type:
Calibrator
Concentration:
50 ng/mL
Validate the Thermo Scientific™ CEDIA™ Mitragynine (Kratom) Assay and ensure accuracy of results using the Thermo Scientific CEDIA Mitragynine (Kratom) Calibrators and Controls (CJ&F). The five calibrators (including negative) establish a standard curve that correlates the instrument signals with the calibrators' respective concentrations in ng/mL. This allows for semi-quantitative measurements. Two controls, one above and one below the cutoff (50ng/mL), validate the calibration curve. The cutoff calibrator provides a reference point for qualitative detection of positive or negative results. For Criminal Justice and Forensic use only in the USA.
  • Ensures assay accuracy
  • Supports qualitative and semi-quantitative measurements
  • Available April 2019
For sale in the U.S.A. only.
For Criminal Justice and Forensic Use Only.
Specifications
Concentration50 ng/mL
Quantity1 x 5 mL
Sample TypeUrine
TypeCalibrator
Detectable AnalytesMitragynine
Unit SizeEach
Contents & Storage
2°C to 8°C