Search
Determine the presence of mitragynine, the major alkaloid found in leaf extracts of the Kratom plant Mitragyna speciosa, in human urine using this automated immunoassay. CJ&F only.
| Catalog Number | Quantity |
|---|---|
| 10026612 | 1 x 65 mL |
| 10026604 | 3 x 17 mL |
Specifically detect mitragynine, the major alkaloid found in leaf extracts of the Kratom plant Mitragyna speciosa, in human urine using this automated immunoassay. For Criminal Justice and Forensic use only in the USA.