Deep Well Plate (96 well) - FAQs
View additional product information for Deep Well Plate (96 well) - FAQs (10045)
1 product FAQs found
Can I concentrate samples during a KingFisher Flex run?
Both deep-well plates and KingFisher 96 plates can be used during the same run. Therefore, it is possible to start the processing by using larger volumes (in a deep-well plate) and elute the purified sample to a smaller volume (in a KingFisher 96 or MagMAX 96 standard plate).