Search
View additional product information for Fetal Bovine Serum, qualified, Australia - FAQs (10099133C)
1 product FAQs found
The main difference is in the Quality Control (QC) test specifications. Qualified and Certified FBS go through the same QC tests but Certified FBS has additional QC tests as well as more stringent QC test specifications for endotoxin and hemoglobin levels. The additional QC tests in Certified FBS include biochemical and hormonal profiling.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.