Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Thermo Scientific™
Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Perform virtually any electrochemistry analysis with these conductivity standards, solutions and accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1010001Calibration Resistor Kit
013017Replacement Flow Cell
Catalog number 1010001
Price (USD)
618.30
Online exclusive
687.00
Save 68.70
Each
Type:
Calibration Resistor Kit
Price (USD)
618.30
Online exclusive
687.00
Save 68.70
Each
Thermo Scientific™Orion&trade Glass Flow Cell is designed for accurate measurement of low-conductivity solutions. When attached directly to the probe, it minimizes the risk of air introduction into the sample during analysis, helping to ensure reliable results.
The Glass Flow Cell is compatible with the Thermo Scientific&tradeOrion&trade 2-Cell Ultrapure Conductivity Probe (Cat. No. 013016MD).
The Thermo Scientific&trade Orion&tradeConductivity Resistor Calibration Kit is designed to verify the performance of Orion&tradeconductivity meters below
Compatible with the following Thermo Scientific™ Orion™ conductivity meters:
  • Versa Star Pro Conductivity Bench Meters (VSTAR20, VSTAR50, VSTAR90, VSTAR91, VSTAR-CND)
  • Pro Star Conductivity Bench Meters (PSTAR2120, PSTAR2150, PSTAR2180)
  • Lab Star Conductivity Bench Meters (LSTAR1120)
  • Legacy Orion Star A210 Conductivity Bench Meters (STARA2120, STARA2150)
  • Legacy Orion Star A110 Conductivity Bench Meters (STARA1120)
Specifications
DescriptionConductivity Calibration Resistor Kit with 8 Pin MiniDIN Connection
For Use With (Application)Conductivity Calibration
For Use With (Equipment)3-Star Conductivity Meter, 4-Star pH/Conductivity Meter, 5-Star pH/DO/Conductivity Meter, 5-Star pH/ISE/DO/Conductivity Meter
TypeCalibration Resistor Kit
Unit SizeEach