Orion™ Star Series RS232 Printer and Computer Cable Set
Orion™ Star Series RS232 Printer and Computer Cable Set
Orion™ Star Series RS232 Printer and Computer Cable Set
Thermo Scientific™
Orion™ Star Series RS232 Printer and Computer Cable Set
Connect your computer and printer to a Versa Star, Star A200 or Star A300 series meter using the Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set.
Have Questions?
Catalog NumberForUseWithEquipment
1010004
Catalog number 1010004
Price (USD)
88.65
Online exclusive
98.50
Save 9.85
Each
Price (USD)
88.65
Online exclusive
98.50
Save 9.85
Each
Connect your computer and printer to a meter using the Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set. These cables are designed for use with Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionOrion Star series RS232 printer and computer cable set
IncludesOne RS232 printer interface cable (2.5mm RS232 meter connector and 25 pin RS232 printer connector) and one RS232 computer interface cable (2.5mm RS232 meter connector and 9 pin RS232 computer connector)
TypeMeter Interface Cable
Unit SizeEach