This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A toRead more
Have Questions?
Change view
Catalog Number
Quantity
101090
also known as 10-1090
1 L
101041
5 mL
101042
25 mL
3 Options
Catalog number 101090
also known as 10-1090
Price (USD)
19,111.65
Online exclusive
20,550.00
Save 1,438.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
19,111.65
Online exclusive
20,550.00
Save 1,438.35
Each
This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.
This product is supplied as a 50% suspension consisting of 25ml Protein A-Sepharose™ 4B and 25 ml of PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.19% Kathon CG/ICP).