Download your free Protein Analysis Guide to protein separation, westerns, ELISAs, and more!
Product Image
Invitrogen™
Rec Protein A
Rec Protein A
Have Questions?
Catalog NumberQuantity
101100
also known as 10-1100
5 mg
Catalog number 101100
also known as 10-1100
Price (USD)
146.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
146.00
Each
This recombinant protein A from Staphylococcus aureus Cowan I was expressed in Bacillus. The gene has been altered to truncate the protein A at the carboxyl terminal. This does not affect the binding characteristics.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeUnconjugated
Quantity5 mg
Shipping ConditionWet Ice
TypeProtein A
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C