Niton™ XL3t XRF Analyzer
Niton™ XL3t XRF Analyzer
Niton™ XL3t XRF Analyzer
Niton™ XL3t XRF Analyzer
Niton™ XL3t XRF Analyzer
Niton™ XL3t XRF Analyzer
Thermo Scientific™

Niton™ XL3t XRF Analyzer

Perform nondestructive testing and get results in seconds for metal analysis, mining and exploration and lead paint testing for toys and consumer goods using handheld analyzers.

Have Questions?
Catalog NumberDetector TypeTube Type
10131166High-performance SemiconductorAu anode 50kV maximum, 200μA maximum
Catalog number 10131166
Price (USD)
-
Detector Type:
High-performance Semiconductor
Tube Type:
Au anode 50kV maximum, 200μA maximum

When you need nondestructive testing with results in seconds for metal analysis, mining and exploration, and lead paint testing for toys and consumer goods, the Thermo Scientific™ Niton™ XL3t XRF Analyzer is the definitive tool for scrap metal recycling, casting and fabrication, manufacturing QA/QC and positive material identification. Get real-time compliance testing, including screening of incoming materials and outgoing finished goods, for the Consumer Product Safety Improvement Act (CPSIA), Proposition 65, Restriction of Hazardous Substances (RoHS) and other regulations.

The feature-leading, handheld Niton XL3t XRF Analyzer offers features engineered to improve your productivity and profitability.

  • High sensitivity and measurement accuracy.
  • Lower detection limits for higher-Z elements, with additional elements Mg, Al, Si and P via helium purge.
  • Standard analytical range: 25 elements from S to U.
  • Optional CCD camera for visually identifying and recording images of samples.
  • A user-selectable 3mm small spot to locate areas of interest on a sample.
  • Flip-up, color, touch-screen display.
  • Ergonomic design.
  • Easy-to-use software.
  • 50 kV x-ray tube provides approximately twice the x-ray flux of a 40 kV x-ray tube.
  • Hotfoot hot surface adaptor allows testing in petrochemical refineries up to 842°F (450°C).

Connectivity

  • Standard Thermo Scientific™ Niton Data Transfer (NDT™) PC software suite allows you to set operator permissions, generate custom reports, print certificates of analysis, or remotely monitor, and operate the instrument hands-free from your PC.
  • Integrated USB and Bluetooth™ communications provide direct data transfer to your PC or networked storage device. Optional accessories include Bluetooth barcode reader, Bluetooth printer, and Bluetooth GPS.
Specifications
Available Analytical ModesVaries by ApplicationAlloy Modes: Metal Alloy, Electronics Alloy, Precious MetalsBulk Modes: Mining, SoilPlastic Modes: RoHS Plastics, Toy and Consumer Goods Plastics, TestAll Painted ProductsOther Modes: Lead Paint, Thin SampleCustom Modes:
Certifications/ComplianceCE, RoHS
Data Memory32Mb internal system memory/ 128Mb internal user storage
DescriptionNiton XL3t XRF Analyzer
Dimensions (L x W x H)9.6 x 9.05 x 3.75 in. (244 x 230 x 95.5 mm)
Display TypeAdjustable angle, color, touch-screen display
Height (English)3.75 in.
Height (Metric)95.5 mm
Length (English)9.6 in.
Length (Metric)244 mm
Max. Power2W
Operating SystemOwnership
Optional AccessoriesPortable test stand, stationary test stand, tripod stand; Extend-a-Pole™ extension pole; Welding Mask; HotFoot™ hot surface adapter; Soil testing guard; Internal CCD sample imaging system; Variable spot size aperture
Power Consumption100 μA
Security FeaturesPassword-protected user security
Standard AccessoriesLocking shielded carrying case, RFID reader, Shielded belt holster, Two 4 (or optional 6) -cell lithium-ion battery packs, Spare battery pack, 110/220 VAC battery charger/AC adaptor, PC connection cables (USB and RS-232), Niton Data Transfer (NDT™) PC software Safety lanyard Check samples/standards
Tube TypeAu anode 50kV maximum, 200μA maximum
TypePortable Analyzer
Voltage50kV
Weight (English)3 lb.
Weight (Metric)1.3kg
Width (English)9.05 in.
Width (Metric)230 mm
ApplicationsElemental Analysis
Detector TypeHigh-performance Semiconductor
Tube Voltage50 kV
Unit SizeEach