Product Image
Thermo Scientific™

Fluorescence Cells

Thermo Scientific™ Fluorescence cells for use with any fluorometer.
Have Questions?
Catalog NumberType
10145334Fluorescence Cell
Catalog number 10145334
Price (USD)
-
Type:
Fluorescence Cell
Thermo Scientific™ Fluorescence cells for use with any fluorometer. Thermo Fisher Scientific offers several standard rectangular, low-volume and test tube cells for fluorescence analysis.
Specifications
For Use With (Equipment)Fluorimeter
TypeFluorescence Cell
Unit SizeEach