Capitol Vial  Confirmation Test Transport Vial
Thermo Scientific™

Capitol Vial Confirmation Test Transport Vial

Thermo Scientific™ Capitol Vial Confirmation Test Transport Vials help confirm test results from on-site test cards or rapid assay cards.
Have Questions?
Catalog NumberVolume (Metric)
10155TS
also known as 10155
45 mL
Catalog number 10155TS
also known as 10155
Price (USD)
-
Volume (Metric):
45 mL

Thermo Scientific™ Capitol Vial Confirmation Test Transport Vials help confirm test results from on-site test cards or rapid assay cards.

  • Tamper evident transport vial 1 oz. (45mL)
  • Ideal choice for on-site testing purposes
  • Helps transport up to 40mL of urine sample to a certified testing laboratory for confirmation testing
Specifications
Description1.52 oz. (45mL) Flip-Top Sample Vial with Tamper Evident Closure
Product TypeTransport Vial
Volume (English)1.5 oz.
Volume (Metric) Total45 mL
Volume (Metric)45 mL
Unit SizeEach