Acetal, 99%
Acetal, 99%
Acetal, 99%
Thermo Scientific Chemicals

Acetal, 99%

CAS: 105-57-7 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 102220010
Price (USD)
212.65
Special offer
Online exclusive
Ends: 28-Dec-2026
250.00
Save 37.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
212.65
Special offer
Online exclusive
Ends: 28-Dec-2026
250.00
Save 37.35
Each
Chemical Identifiers
CAS105-57-7
IUPAC Name1,1-diethoxyethane
Molecular FormulaC6H14O2
InChI KeyDHKHKXVYLBGOIT-UHFFFAOYSA-N
SMILESCCOC(C)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Density(20°C) 0.822 to 0.829 kg/l
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only