Acetoacetanilide, 99%
Acetoacetanilide, 99%
Acetoacetanilide, 99%
Thermo Scientific Chemicals

Acetoacetanilide, 99%

CAS: 102-01-2 | C10H11NO2 | 177.2 g/mol
Have Questions?
Catalog NumberQuantity
1023600505 g
Catalog number 102360050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS102-01-2
IUPAC Name3-oxo-N-phenylbutanamide
Molecular FormulaC10H11NO2
InChI KeyDYRDKSSFIWVSNM-UHFFFAOYSA-N
SMILESCC(=O)CC(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)White
Appearance (Form)Powder or crystals
Melting point83°C to 86°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only