Acetone oxime, 98%
Acetone oxime, 98%
Acetone oxime, 98%
Thermo Scientific Chemicals

Acetone oxime, 98%

CAS: 127-06-0 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 102395000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS127-06-0
IUPAC NameN-(propan-2-ylidene)hydroxylamine
Molecular FormulaC3H7NO
InChI KeyPXAJQJMDEXJWFB-UHFFFAOYSA-N
SMILESCC(C)=NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
GC>=97.5 %
Appearance (Form)Crystals or crystalline solid
Melting point58°C to 63°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only