3-Acetylpyridine, 98%
3-Acetylpyridine, 98%
3-Acetylpyridine, 98%
Thermo Scientific Chemicals

3-Acetylpyridine, 98%

CAS: 350-03-8 | C7H7NO | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
500 g
Packaging:
Glass bottle
Catalog number 102690100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS350-03-8
IUPAC Name1-(pyridin-3-yl)ethan-1-one
Molecular FormulaC7H7NO
InChI KeyWEGYGNROSJDEIW-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5330 to 1.5370 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only