2-Acetylthiophene, 98%
2-Acetylthiophene, 98%
2-Acetylthiophene, 98%
Thermo Scientific Chemicals

2-Acetylthiophene, 98%

CAS: 88-15-3 | C6H6OS | 126.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 102721000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-15-3
IUPAC Name1-(thiophen-2-yl)ethan-1-one
Molecular FormulaC6H6OS
InChI KeyWYJOVVXUZNRJQY-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear light yellow to light brown
Refractive index1.5640 to 1.5660 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only