N-Allylaniline, 95%
N-Allylaniline, 95%
N-Allylaniline, 95%
Thermo Scientific Chemicals

N-Allylaniline, 95%

CAS: 589-09-3 | C9H11N | 133.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number 102870050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS589-09-3
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5620 to 1.5660 (20°C, 589 nm)
Appearance (Color)Clear yellow to orange to brown
Appearance (Form)Liquid
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only