Allylbenzene, 98%
Allylbenzene, 98%
Allylbenzene, 98%
Thermo Scientific Chemicals

Allylbenzene, 98%

CAS: 300-57-2 | C9H10 | 118.18 g/mol
数量:
25 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 102891000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS300-57-2
IUPAC Name(prop-2-en-1-yl)benzene
Molecular FormulaC9H10
InChI KeyHJWLCRVIBGQPNF-UHFFFAOYSA-N
SMILESC=CCC1=CC=CC=C1
さらに表示
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.5095 to 1.5135 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only