3-Amino-4-carbethoxypyrazole, 99%
3-Amino-4-carbethoxypyrazole, 99%
3-Amino-4-carbethoxypyrazole, 99%
Thermo Scientific Chemicals

3-Amino-4-carbethoxypyrazole, 99%

CAS: 6994-25-8 | C6H9N3O2 | 155.16 g/mol
Have Questions?
Catalog number 103310050
Price (USD)
65.30
Each
Request bulk or custom format
Price (USD)
65.30
Each
Chemical Identifiers
CAS6994-25-8
IUPAC Nameethyl 5-amino-1H-pyrazole-4-carboxylate
Molecular FormulaC6H9N3O2
InChI KeyYPXGHKWOJXQLQU-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(N)NN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Melting point103°C to 107°C
Appearance (Color)White to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only