2-Amino-4-chlorobenzoic acid, 98%
2-Amino-4-chlorobenzoic acid, 98%
2-Amino-4-chlorobenzoic acid, 98%
Thermo Scientific Chemicals

2-Amino-4-chlorobenzoic acid, 98%

CAS: 89-77-0 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 103321000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS89-77-0
IUPAC Name2-amino-4-chlorobenzoic acid
Molecular FormulaC7H6ClNO2
InChI KeyJYYLQSCZISREGY-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Powder
Melting point231°C to 235°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only