2-Amino-4,6-dichloropyrimidine, 95%, tech.
2-Amino-4,6-dichloropyrimidine, 95%, tech.
2-Amino-4,6-dichloropyrimidine, 95%, tech.
Thermo Scientific Chemicals

2-Amino-4,6-dichloropyrimidine, 95%, tech.

CAS: 56-05-3 | C4H3Cl2N3 | 163.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 103490500
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS56-05-3
IUPAC Name4,6-dichloropyrimidin-2-amine
Molecular FormulaC4H3Cl2N3
InChI KeyJPZOAVGMSDSWSW-UHFFFAOYSA-N
SMILESNC1=NC(Cl)=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point216°C to 226°C
Infrared spectrumConforms
HPLC>=94 %
Appearance (Color)Yellow to orange to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only