2-Amino-4,6-dihydroxypyrimidine, 98%
2-Amino-4,6-dihydroxypyrimidine, 98%
2-Amino-4,6-dihydroxypyrimidine, 98%
Thermo Scientific Chemicals

2-Amino-4,6-dihydroxypyrimidine, 98%

CAS: 56-09-7 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 103510250
Price (EUR)
46,90
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
46,90
Each
Chemical Identifiers
CAS56-09-7
IUPAC Name2-amino-6-hydroxy-3,4-dihydropyrimidin-4-one
Molecular FormulaC4H5N3O2
InChI KeyAUFJTVGCSJNQIF-UHFFFAOYSA-N
SMILESNC1=NC(O)=CC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Titration excess NaOH>=97.5 %
Appearance (Color)Off-white to light yellow to light pink
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only