2-(2-Aminoethoxy)ethanol, 98%
2-(2-Aminoethoxy)ethanol, 98%
2-(2-Aminoethoxy)ethanol, 98%
Thermo Scientific Chemicals

2-(2-Aminoethoxy)ethanol, 98%

CAS: 929-06-6 | C4H11NO2 | 105.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 103630025
Price (USD)
336.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
336.00
Each
Chemical Identifiers
CAS929-06-6
IUPAC Name2-(2-aminoethoxy)ethan-1-ol
Molecular FormulaC4H11NO2
InChI KeyGIAFURWZWWWBQT-UHFFFAOYSA-N
SMILESNCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4570 to 1.4610 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only