2-(2-Aminoethyl)pyridine, 95%
2-(2-Aminoethyl)pyridine, 95%
2-(2-Aminoethyl)pyridine, 95%
Thermo Scientific Chemicals

2-(2-Aminoethyl)pyridine, 95%

CAS: 2706-56-1 | C7H10N2 | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 103690100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2706-56-1
IUPAC Name2-(pyridin-2-yl)ethan-1-amine
Molecular FormulaC7H10N2
InChI KeyXPQIPUZPSLAZDV-UHFFFAOYSA-N
SMILESNCCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5340 to 1.5380 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only