2-Aminoindan hydrochloride, 98%
2-Aminoindan hydrochloride, 98%
2-Aminoindan hydrochloride, 98%
Thermo Scientific Chemicals

2-Aminoindan hydrochloride, 98%

CAS: 2338-18-3 | C9H11ClN | 168.64 g/mol
数量:
1 g
5 g
25 g
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 103860050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2338-18-3
IUPAC Name2,3-dihydro-1H-inden-2-amine hydrochloridyl
Molecular FormulaC9H11ClN
InChI KeyDLYRFJDJKGGVKT-UHFFFAOYSA-N
SMILES[Cl].NC1CC2=CC=CC=C2C1
さらに表示
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only