5-Amino-2-methoxypyridine, 90%, tech.
5-Amino-2-methoxypyridine, 90%, tech.
5-Amino-2-methoxypyridine, 90%, tech.
Thermo Scientific Chemicals

5-Amino-2-methoxypyridine, 90%, tech.

CAS: 6628-77-9 | C6H8N2O | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 103950250
Price (USD)
148.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
148.00
Each
Chemical Identifiers
CAS6628-77-9
IUPAC Name6-methoxypyridin-3-amine
Molecular FormulaC6H8N2O
InChI KeyUUVDJIWRSIJEBS-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Red to yellow-brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only