2-Amino-4-methylpyrimidine, 97%
2-Amino-4-methylpyrimidine, 97%
2-Amino-4-methylpyrimidine, 97%
Thermo Scientific Chemicals

2-Amino-4-methylpyrimidine, 97%

CAS: 108-52-1 | C5H7N3 | 109.132 g/mol
数量:
10 g
100 g
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 104091000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-52-1
IUPAC Name4-methylpyrimidin-2-amine
Molecular FormulaC5H7N3
InChI KeyGHCFWKFREBNSPC-UHFFFAOYSA-N
SMILESCC1=CC=NC(N)=N1
さらに表示
Appearance (Color)Off-white to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point155°C to 160°C
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only