3-Amino-2-naphthoic acid, 85%, tech.
3-Amino-2-naphthoic acid, 85%, tech.
3-Amino-2-naphthoic acid, 85%, tech.
Thermo Scientific Chemicals

3-Amino-2-naphthoic acid, 85%, tech.

CAS: 5959-52-4 | C11H9NO2 | 187.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 104140050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5959-52-4
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=80 %
Appearance (Form)Powder
HPLC>=80 %
Infrared spectrumConforms
Appearance (Color)Yellow to green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only