(±)-2-Amino-1-phenylethanol, 95%
(±)-2-Amino-1-phenylethanol, 95%
(±)-2-Amino-1-phenylethanol, 95%
Thermo Scientific Chemicals

(±)-2-Amino-1-phenylethanol, 95%

CAS: 7568-93-6 | C8H11NO | 137.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
10432010010 gGlass bottle
Catalog number 104320100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7568-93-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only