1-Aminopiperidine, 97%
1-Aminopiperidine, 97%
1-Aminopiperidine, 97%
Thermo Scientific Chemicals

1-Aminopiperidine, 97%

CAS: 2213-43-6 | C5H12N2 | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 104410100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2213-43-6
IUPAC Namepiperidin-1-amine
Molecular FormulaC5H12N2
InChI KeyLWMPFIOTEAXAGV-UHFFFAOYSA-N
SMILESNN1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4735 to 1.4775 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only