3-Amino-1,2-propanediol, 98%
3-Amino-1,2-propanediol, 98%
3-Amino-1,2-propanediol, 98%
Thermo Scientific Chemicals

3-Amino-1,2-propanediol, 98%

CAS: 616-30-8 | C3H9NO2 | 91.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 104420250
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.70
Save 13.05
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.70
Save 13.05
Each
Chemical Identifiers
CAS616-30-8
IUPAC Name3-aminopropane-1,2-diol
Molecular FormulaC3H9NO2
InChI KeyKQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILESNCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Water=<0.5 % (K.F.)
Refractive index1.4900 to 1.4940 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only